Anti-Smad2 Rabbit Polyclonal Antibody [clone: Y216]

Supplier: Bioworld Technology
BS1769
BWRLBS1769EA 283 EUR
BWRLBS1769
Anti-Smad2 Rabbit Polyclonal Antibody [clone: Y216]
Antibodies
Synthetic peptide, corresponding to amino acids 185-234 of Human Smad2.

Smad2 (Y216) pAb detects endogenous levels of Smad2 protein.

Type: Primary
Antigen: Smad2
Clonality: Polyclonal
Clone: Y216
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

SPECIFICATIONS

Antigen symbol SMAD family member 2
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Clone Y216
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR