Anti-TFIIH p62 Rabbit Polyclonal Antibody [clone: A32]

Supplier: Bioworld Technology
BS2271
BWRLBS2271EA 283 EUR
BWRLBS2271
Anti-TFIIH p62 Rabbit Polyclonal Antibody [clone: A32]
Antibodies
Synthetic peptide, corresponding to amino acids N-terminus of Human TFIIH p62.

TFIIH p62 (A32) pAb detects endogenous levels of TFIIH p62 protein.

Type: Primary
Antigen: TFIIH p62
Clonality: Polyclonal
Clone: A32
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR