Anti-Casein Kinase Iepsilon Rabbit Polyclonal Antibody [clone: P301]

Supplier: Bioworld Technology
BS2279
BWRLBS2279EA 283 EUR
BWRLBS2279
Anti-Casein Kinase Iepsilon Rabbit Polyclonal Antibody [clone: P301]
Antibodies
Synthetic peptide, corresponding to amino acids 250-350 of Human Casein Kinase Iepsilon.

Casein Kinase I? (P301) pAb detects endogenous levels of Casein Kinase I? protein.

Type: Primary
Antigen: Casein Kinase Iepsilon
Clonality: Polyclonal
Clone: P301
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR