Anti-p-NPM Rabbit Polyclonal Antibody [clone: T234]

Supplier: Bioworld Technology
BS4656
BWRLBS4656EA 307 EUR
BWRLBS4656
Anti-p-NPM Rabbit Polyclonal Antibody [clone: T234]
Antibodies
Synthetic phosphopeptide derived from human NPM around the phosphorylation site of Threonine 234.

p-NPM (T234) pAb detects endogenous levels of NPM protein only when phosphorylated at Thr234.

Type: Primary
Antigen: p-NPM
Clonality: Polyclonal
Clone: T234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

SPECIFICATIONS

Antigen symbol p-NPM
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Clone T234
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR