Anti-IQGAP1 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
BS5767
BWRLBS5767EA 283 EUR
BWRLBS5767
Anti-IQGAP1 Rabbit Polyclonal Antibody
Antibodies
Synthetic peptide, corresponding to amino acids 291-336 of Human IQGAP1.

IQGAP1 pAb detects endogenous levels of IQGAP1 protein.

Type: Primary
Antigen: IQGAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR