Anti-SLC8A1 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
BS5816
BWRLBS5816EA 283 EUR
BWRLBS5816
Anti-SLC8A1 Rabbit Polyclonal Antibody
Antibodies
Synthetic peptide, corresponding to amino acids 336-381 of Human SLC8A1.

SLC8A1 pAb detects endogenous levels of SLC8A1 protein.

Type: Primary
Antigen: SLC8A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR