Multi-gas detectors, GasAlert Quattro
Supplier: BW TECHNOLOGIES
QT-XW00-R-Y-EU QT-XW0M-R-Y-EU QT-XWH0-R-Y-EU QT-XWHM-R-Y-EU QT-XWHM-A-Y-EU QT-X00M-A-Y-EU QT-X00M-R-Y-EU QT-00HM-A-Y-EU QT-00HM-R-Y-EU QT-000M-R-Y-EU QT-00H0-R-Y-EU QT-00H0-A-Y-EU QT-000M-A-Y-EU QT-X0HM-R-Y-EU QT-XW0M-A-Y-EU QT-XW00-R-Y-UK QT-000M-A-Y-UK QT-000M-R-Y-UK QT-00H0-A-Y-UK QT-00H0-R-Y-UK QT-00HM-A-Y-UK QT-00HM-R-Y-UK QT-X00M-A-Y-UK QT-X00M-R-Y-UK QT-X0HM-R-Y-UK QT-XW0M-A-Y-UK QT-XW0M-R-Y-UK QT-XWH0-R-Y-UK QT-XWHM-R-Y-UK
BWTEQT-XW00-R-Y-EUEA 591 EUR
BWTEQT-XW00-R-Y-EU BWTEQT-XW0M-R-Y-EU BWTEQT-XWH0-R-Y-EU BWTEQT-XWHM-R-Y-EU BWTEQT-XWHM-A-Y-EU BWTEQT-X00M-A-Y-EU BWTEQT-X00M-R-Y-EU BWTEQT-00HM-A-Y-EU BWTEQT-00HM-R-Y-EU BWTEQT-000M-R-Y-EU BWTEQT-00H0-R-Y-EU BWTEQT-00H0-A-Y-EU BWTEQT-000M-A-Y-EU BWTEQT-X0HM-R-Y-EU BWTEQT-XW0M-A-Y-EU BWTEQT-XW00-R-Y-UK BWTEQT-000M-A-Y-UK BWTEQT-000M-R-Y-UK BWTEQT-00H0-A-Y-UK BWTEQT-00H0-R-Y-UK BWTEQT-00HM-A-Y-UK BWTEQT-00HM-R-Y-UK BWTEQT-X00M-A-Y-UK BWTEQT-X00M-R-Y-UK BWTEQT-X0HM-R-Y-UK BWTEQT-XW0M-A-Y-UK BWTEQT-XW0M-R-Y-UK BWTEQT-XWH0-R-Y-UK BWTEQT-XWHM-R-Y-UK
Multi-gas detectors, GasAlert Quattro
Gas Detection Systems
The rugged GasAlert Quattro reliably monitors up to four gas and combines a one-button operation with flexible options.
  • Compatible with BW MicroDock II automatic test and calibration station
  • Graphic LCD displays to easily identify icons
  • IntelliFlash provides continuous visual confirmation of detector operation and compliance
Certifications: ACGIH 1 ppm H2S TWA recommendations.
Delivery information: Supplied with a rechargeable battery pack or alkaline pack with three AA batteries, charger wall plug (with rechargeable battery option), calibrationMore Product Information