Cyclohexane ≥99.8%, Laboratory Reagent

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

C100307-10L C100307-2.5L C100307-25L C100307-1L C100307-4X2.5L
C100307-10LEA 470 EUR
C100307-10L C100307-2.5L C100307-25L C100307-1L C100307-4X2.5L
Cyclohexane ≥99.8%, Laboratory Reagent
Cyclohexane

  • ≥95% for general laboratory use.


Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 8.3 %(V)
Melting Pt: 6.4 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (form) complying
Appearance (color) complying
Purity (GC) Min. 99.80 Fl.%
Non-volatile matter Max. 0.005 %
IR spectrum complying
Vapor Density 2.9 (vs air)
Vapor Pressure 77 mmHg ( 20 °C)
Autoignition Temperature 500 °F
Refractive Index n20/D 1.426 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR