Estriol

Supplier: Cayman Chemical

10006484-100 10006484-500 10006484-1 10006484-5
CAYM10006484-100EA 23.2 EUR
CAYM10006484-100 CAYM10006484-500 CAYM10006484-1 CAYM10006484-5
Estriol
Estriol
Formula: C₁₈H₂₄O₃
MDL Number: MFCD00003691
CAS Number: 50-27-1
EINECS: 200-022-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR