(R)-(-)-β-Aminophenethyl alcohol

Supplier: Cayman Chemical

10007132-1 10007132-10 10007132-25 10007132-5
CAYM10007132-1EA 34.9 EUR
CAYM10007132-1 CAYM10007132-10 CAYM10007132-25 CAYM10007132-5
(R)-(-)-β-Aminophenethyl alcohol
(R)-(-)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR