Anti-SPHK 1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Cayman Chemical
10012201-500
CAYM10012201-500EA 570 EUR
CAYM10012201-500
Anti-SPHK 1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: SPHK 1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR