4-Methoxythiobenzamide

Supplier: Cayman Chemical

10552-500 10552-1 10552-5
CAYM10552-500EA 34.2 EUR
CAYM10552-500 CAYM10552-1 CAYM10552-5
4-Methoxythiobenzamide
4-Methoxythiobenzamide
Formula: C₈H₉NOS
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 155 °C
MDL Number: MFCD00040993
CAS Number: 2362-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR