Anti-Syn1 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
10606-1
CAYM10606-1EA 787 EUR
CAYM10606-1
Anti-Syn1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Syn1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR