3-Aminothiobenzamide

Supplier: Cayman Chemical

11064-500 11064-1 11064-5
CAYM11064-500EA 53.7 EUR
CAYM11064-500 CAYM11064-1 CAYM11064-5
3-Aminothiobenzamide
3-Aminothiobenzamide
Formula: C₇H₈N₂S
MDL Number: MFCD04973325
CAS Number: 78950-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR