Cytochalasin D

Supplier: Cayman Chemical

11330-1 11330-5 11330-10
CAYM11330-1EA 101 EUR
CAYM11330-1 CAYM11330-5 CAYM11330-10
Cytochalasin D
Cytochalasin D
Formula: C₃₀H₃₇NO₆
Melting Pt: 259…260 °C
MDL Number: MFCD00077706
CAS Number: 22144-77-0
EINECS: 244-804-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR