Thioisonicotinamide

Supplier: Cayman Chemical

13056-1 13056-5 13056-10 13056-25
CAYM13056-1EA 19.6 EUR
CAYM13056-1 CAYM13056-5 CAYM13056-10 CAYM13056-25
Thioisonicotinamide
Thioisonicotinamide
Formula: C₆H₆N₂S
MDL Number: MFCD00006437
CAS Number: 2196-13-6
EINECS: 218-592-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR