Melatonin

Supplier: Cayman Chemical

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

14427-5 14427-10
CAYM14427-5EA 52.5 EUR
CAYM14427-5 CAYM14427-10
Melatonin
Melatonine
Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR