(-)-Indolactam V ≥97%, solid

Supplier: Cayman Chemical

14647-1 14647-500
CAYM14647-1EA 596 EUR
CAYM14647-1 CAYM14647-500
(-)-Indolactam V ≥97%, solid
(-)-Indolactam V

(−)-Indolactam V is a tumor-promoting teleocidin compound that activates protein kinase C (PKC). It binds to PKC regulatory domains of mouse skin PKCη and rat brain PKCγ with Ki values of 3,4 nM and 1 μM, respectively. At 1 μM, (−)-indolactam V induces differentiation of human embryonic stem cells into pancreatic progenitors through activation of PKC signaling.

Formula: C₁₇H₂₃N₃O₂
MW: 301.39 g/mol
CAS Number: 90365-57-4

Order Now

Specification Test Results

Formal name (2S,5S)-1,2,4,5,6,8-hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3H-pyrrolo[4,3,2-gh]-1,4-benzodiazonin-3-one
CAS number 90365-57-4
Molecular formula C17H23N3O2
Formula weight 301,4
Purity ≥97%
Formulation A white to off-white solid
λmax 203, 229 and 301 nm
Storage 4 °C
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR