Anti-Smt3 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
14697-1
CAYM14697-1EA 510 EUR
CAYM14697-1
Anti-Smt3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Smt3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR