Calcein fluorescent indicator

Supplier: Cayman Chemical
Warning

16221-1 16221-25 16221-5 16221-10
CAYM16221-1EA 52.5 EUR
CAYM16221-1 CAYM16221-25 CAYM16221-5 CAYM16221-10
Calcein fluorescent indicator
Calcein

Calcein is a xanthene that is commonly used for the fluorometric determination of calcium in solution. As this form of calcein is not membrane permeable, it can be used in assays that evaluate membrane integrity. Calcein can also be used as a fluorescent indicator for fluoride, iron, and mercury. Excitation and emission maxima for calcein are 494 and 517 nm, respectively.


  • Emission 517 nm
  • Excitation 494 nm
  • Formulation A crystalline solid
  • λmax 226, 278 nm


This form of calcein is not membrane permeable and can be used in assays that evaluate membrane integrity.

Formula: C₃₀H₂₆N₂O₁₃
Storage Temperature: Freezer
MDL Number: MFCD00005049
CAS Number: 1461-15-0
EINECS: 215-957-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR