Anti-CD36 Mouse Monoclonal Antibody [clone: JC63.1]

Supplier: Cayman Chemical
188150-100
CAYM188150-100EA 510 EUR
CAYM188150-100
Anti-CD36 Mouse Monoclonal Antibody [clone: JC63.1]
Antibodies
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: JC63.1
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgA
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR