1-(3,4-Dihydroxyphenyl)propan-2-one ≥98%, solid

Supplier: Cayman Chemical
Warning

19522-25
CAYM19522-25EA 152 EUR
CAYM19522-25
1-(3,4-Dihydroxyphenyl)propan-2-one ≥98%, solid
1-(3,4-Dihydroxyphenyl)propan-2-one

3',4'-Dihydroxyphenylacetone (Item No. 19522) is an analytical reference standard. It is a minor metabolite of 3,4-MDEA (Item Nos. 14085 | 15689), MDMA, and α-methyldopa produced by oxidative deamination.1,2,3 This product is intended for forensic and research applications.


Caution: This product is not for human or veterinary use.

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD03093024
CAS Number: 2503-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR