Cysteaminium chloride ≥95%, crystalline solid

Supplier: Cayman Chemical
Danger

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

22193-10 22193-100 22193-25 22193-50
CAYM22193-10EA 52.5 EUR
CAYM22193-10 CAYM22193-100 CAYM22193-25 CAYM22193-50
Cysteaminium chloride ≥95%, crystalline solid
Cysteaminium chloride

Cysteamine is a stable aminothiol with radioprotective activities.


It reduces ionizing radiation-induced death and chromosomal damage in mice in a dose-dependent manner. Cysteamine binds rapidly and temporarily to plasma proteins upon administration and this activity is directly correlated to its radioprotective effects. In vitro, 0.1 mM cysteamine depletes 90% of free cystine from cystinotic fibroblasts. Formulations containing cysteamine have been used to treat nephropathic cystinosis and reduce glomerular deterioration in humans.


Caution: This product is not for human or veterinary use.

Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012904
CAS Number: 156-57-0
UN: 3335
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR