Long-Chain Cis/Trans Isomer Fatty Acid Methyl Ester Mixture
Supplier: Cayman Chemical
29363-5
CAYM29363-5EA 337 EUR
CAYM29363-5
Long-Chain Cis/Trans Isomer Fatty Acid Methyl Ester Mixture
Standards Organic Standards
Long-chain cis/trans isomer fatty acid methyl ester mixture contains cis and trans isomers of several fatty acids. It is intended for use as a reference standard for the detection of long-chain fatty acids in various sample types by GC-MS.
  • A solution in dichloromethane
  • Stability: ≥1 year
  • GC Column used: SP-2560
Packaging: Package includes with C16:0, C18:0, C:18:1 (6-trans, 8-trans), C:18:1 (9-trans), C:18:1 (10-trans), C:18:1 (11-trans), C:18:1 (12-trans), C:18:1 (13-trans, 14-trans, 6-cis, 8-cis), C:18:1 (9-cis), C:18:1 (15-trans, 10-cis), C:18:1 (11-cis), C:18:1 (12-cis), C18:2, C20:0, C18:3, C22:0.