BSA-Oleate Monounsaturated Fatty Acid Complex

Supplier: Cayman Chemical

29557-5
CAYM29557-5EA 248 EUR
CAYM29557-5
BSA-Oleate Monounsaturated Fatty Acid Complex
Standards Food Standards

BSA-Oleate monounsaturated fatty acid complex is composed of oleic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of oleate:BSA. It has been used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation or similar processes in various cellular metabolic studies.


  • A fatty acid substrate
  • Origin: Animal/Bovine
  • Stability: ≥ 1 year


The BSA/BSA-FAs are acceptable for use with short-term cell culture applications (acute treatment to 18 hours), however for long-term applications (25+ hours) the product can/should be filter sterilized using a 0.2 µm filter and sterile receptacle without affecting the performance. For best results, it is recommended that this product be used in conjunction with BSA control for fatty acid complexes (5 mM).


Formulation: 5 mM Oleate:0.8 mM BSA (6:1 oleate:BSA) in 150 mM sodium chloride, pH 7.4


Caution: Store at –20 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR