Cell-based assay calcein AM
Supplier: Cayman Chemical
400146-1
CAYM400146-1EA 164 EUR
CAYM400146-1
Cell-based assay calcein AM
Assays Cellular Assays
Packaging: This vial contains 50 µl of 1 mM calcein AM solution in DMSO.