Fluprostenol ELISA kit
Supplier: Cayman Chemical
516761-480 516761-96 516761-96S 516761-480S
CAYM516761-480EA 1530 EUR
CAYM516761-480 CAYM516761-96 CAYM516761-96S CAYM516761-480S
Fluprostenol ELISA kit
Assays ELISAs
A sensitive detection method for measuring the free acid form of fluprostenol.
  • Measure the free acid form of fluprostenol
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure fluprostenol levels down to 16 pg/ml
  • Colorimetric reading at 405 to 420 nm
Fluprostenol is a metabolically stable analog of PGF with potent FP receptor agonist activity. The isopropyl ester of fluprostenol has recently been approved for use as an ocular hypotensive drug for the treatment of glaucoma and is sold under the Alcon trade name Travoprost. Acting as a prodrug, Fluprostenol isopropyl ester is converted by esterase activity in the cornea to yield the corresponding active free acid, (+)-fluprostenol. Fluprostenol is also used in the veterinary field for the induction of estrus in livestock. Cayman’s Fluprostenol ELISA is a sensitive detection method for measuring the free acid form of fluprostenol. The assay also detects the isopropyl ester form of fluprostenol, but with less sensitivity. Samples containing a mixture of the isopropyl ester and free acid should be purified and the compounds separated prior to analysis. The assay has been validated for use in aqueous humor samples.
Delivery information: kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent.