DEA NONOate ≥98%, crystalline solid

Supplier: Cayman Chemical
Danger

Synonyms: 2-(N,N-diethylamino)diazenolate-2-oxide diethylammonium salt

82100-10 82100-100 82100-50 82100-500
CAYM82100-10EA 41.5 EUR
CAYM82100-10 CAYM82100-100 CAYM82100-50 CAYM82100-500
DEA NONOate ≥98%, crystalline solid
DEA NONOate

DEA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 2 minutes and 16 minutes at 37 °C and 22 to 25 °C, pH 7.4, respectively, to liberate 1.5 moles of NO per mole of parent compound.


Caution: Not for human or veterinary use.

Formula: C₄H₁₀N₃O₂·C₄H₁₂N
CAS Number: 372965-00-9

Order Now

Specification Test Results

Purity ≥98%
λmax 250 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR