Alkaline detergent, RBS IND 470
Supplier: CHEMICAL
RBSIND470
CHEPRBSIND470EA 111 EUR
CHEPRBSIND470
Alkaline detergent, RBS IND 470
Cleaning Agents Detergents and Surface Cleansers
Concentrated alkaline liquid detergent for radioactive decontamination of parts and surfaces made of glass and stainless steel.
  • Cleans and decontaminates
  • Halogen-free
  • high emulsifying and detergency power
RBS IND 470 is recommended for manual cleaning and soaking of parts, surfaces and containers made of stainless steel, glass and plastics. It is suitable for aqueous cleaning processes in laboratories and industry (nuclear power station, cyclotrons and others institutions that use to handle radioactive products).
Ordering information: Available inMore Product Information