Anti-T7 Tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
D3-1832
COBSD3-1832EA 629 EUR
COBSD3-1832
Anti-T7 Tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Tag
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR