Mouse IgG2b Isotype Control (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-1602B
COBSD5-1602BEA 385 EUR
COBSD5-1602B
Mouse IgG2b Isotype Control (RPE (R-Phycoerythrin))
Antibodies
Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: None
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR