Anti-c-Myc tag Mouse Monoclonal Antibody SureLight® P3 [clone: 9E10]

Supplier: Columbia Biosciences
D7-1714
COBSD7-1714EA 629 EUR
COBSD7-1714
Anti-c-Myc tag Mouse Monoclonal Antibody SureLight® P3 [clone: 9E10]
Antibodies
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: SureLight® P3
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR