Anti-6X His tag Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: AD1.1.10]

Supplier: Columbia Biosciences
HRP-1711
COBSHRP-1711EA 629 EUR
COBSHRP-1711
Anti-6X His tag Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: AD1.1.10]
Antibodies
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR