Needles, STERITEX®
Supplier: CODAN
646008
CODA646008EA 134 EUR
CODA646008
Needles, STERITEX®
Syringe Needles
This needle is only used for the inlet of air into bottles.
  • Metal needle with non return valve
  • With 0,2 µm air filter
Caution: Do not use for reconstitution of lyophilisate.