Stainless Steel Buckets with Spout
Supplier: Adelphi Pharma Hygiene Products
BUL4010P BUL4012P BUL4015P
COLDBUL4010PEA 94.9 EUR
COLDBUL4010P COLDBUL4012P COLDBUL4015P
Stainless Steel Buckets with Spout
Buckets
Hygienic, high-quality stainless steel buckets meet international pharma quality compliance regulations. Available in 304 grade stainless steel, these buckets are bright polished throughout, graduated in 1 L intervals, and come complete with a swing handle.
  • Buckets available as standard, off-the-shelf for delivery within days
  • Buckets in 304 grade stainless steel
  • Optional lids available to purchase separately
  • Exceptional average surface finish of 0,4 µm Ra
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable