VWR® ACE® UltraCore Super C18 HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
CORE-25A-15005EA 1120 EUR
CORE-25A-15005 CORE-25A-10005 CORE-25A-0301 CORE-25A-03005 CORE-25A-7546U CORE-25A-7503U CORE-25A-7502U CORE-25A-3503U CORE-25A-1546U CORE-25A-1503U CORE-25A-1502U CORE-25A-1246U CORE-25A-1203U CORE-25A-1202U CORE-25A-1046U CORE-25A-1003U CORE-25A-1002U CORE-25A-0546U CORE-25A-0503U CORE-25A-0502U CORE-25A-0346U CORE-25A-0303U CORE-25A-0302U CORE-25A-0246U CORE-25A-0203U CORE-25A-0202U CORE-A-GD2U CORE-A-GD4U
VWR® ACE® UltraCore Super C18 HPLC/UHPLC Columns, 2,5 µm
Chromatography Columns
Avantor® ACE® UltraCore SuperC18 solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These stainless steel columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability.
  • Ideal for demanding MS applications
More Product Information