Magnetic stirrers, Corning® PC-210
Supplier: Corning
6796-210
CORN6796-210EA 433 EUR
CORN6796-210
Magnetic stirrers, Corning® PC-210
Stirrers Magnetic Stirrers
The Corning® stirrers have a durable glass-ceramic 4×5" Pyroceram® top and offer superior performance and reliability in a low profile, space-saving design.
  • Designed with CEE7-7 plug
  • Microprocessor controlled to automatically adjust motor strength to any solution
  • Enclosed electronics design protects components, improves performance, and prolongs product life
  • Two piece casting design deflects spills away from electronics
  • Built in mount for an optional support rod
Exclusive closed-loop stirring control monitors and regulates the stirring speed forMore Product Information