Supelco® TraceCERT® Organic Reference Standard, Diclofop-methyl
Supplier: Merck
CRM04291-100MG
CRM04291-100MGEA 121 EUR
CRM04291-100MG
Supelco® TraceCERT® Organic Reference Standard, Diclofop-methyl
Standards Organic Standards
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The diclofop-methyl CRM may find its use as described below:
  • To study the enantioselective degradation of diclofop-methyl and diclofop in two soil samples under aerobic and anaerobic conditions using high-performance liquid chromatography (HPLC)
  • Simultaneous extraction and determination of enantiomers of diclofop-methyl and diclofop in cole using high-performance liquid chromatography-chiral stationary phase (HPLC-CS)
  • Development of a multi-residue method for simultaneous estimation of 16 pesticides in water samples using direct immersion SPME followed by GC-MS analysis
  • Simultaneous extraction and determination of aryloxyphenoxy-propionate herbicides from water samples by dispersive magnetic solid-phase extraction (d-MSPE) with high-performance liquid chromatography-diode array detector (HPLC-DAD) and ultra-high pressure liquid chromatography -triple quadrupole mass spectrometer (UHPLC-MS/MS)
  • Evaluating the capacity of silica-bonded deoxycholic acid as a stationary phase having a 3,5-dinitrophenylcarbamoyl and a calix[4]arene substituent for enantiomeric separations
Caution: P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352