Supelco® TraceCERT® Organic Reference Standard, Disulfoton-sulfoxide
Supplier: Merck
CRM05207-50MG
CRM05207-50MGEA 143 EUR
CRM05207-50MG
Supelco® TraceCERT® Organic Reference Standard, Disulfoton-sulfoxide
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312