Dibucaine hydrochloride, >=99%, analytical standard, Supelco®

Supplier: Merck

D0638-5G D0638-1G D0638-25G
D0638-5GEA 158 EUR
D0638-5G D0638-1G D0638-25G
Dibucaine hydrochloride, >=99%, analytical standard, Supelco®
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b20f10a1d0a3672e12c668811f8200484e176445&vid=ce7621d3ac647aa4f9f5d99e3ddd0c1c1ee1973f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=988ba3204ccae04f9c89823e699bcf194c0d1cd9&vid=89d31e94b21bc01d784623072c4a5aaea353bec5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7b1422606da256e2e33f18ce31952a672061686&vid=4a16e6a4459de7595ea1b3d5c8bcb52ee4250702&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Description Dibucaine hydrochloride Dibucaine hydrochloride
M-Clarity quality level MQ200
Environmentally Preferable
CAS No. 61-12-1 61-12-1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR