Cut resistant gloves, ChainExtra, with short cuff
Supplier: HONEYWELL SAFETY
2542411R0302 2542412R0302 2542413R0302 2542414R0302
DALL2542411R0302EA 331 EUR
DALL2542411R0302 DALL2542412R0302 DALL2542413R0302 DALL2542414R0302
Cut resistant gloves, ChainExtra, with short cuff
Gloves
These fully reversible chainmail gloves are equipped with heavy duty adjustable straps and press stud fastening. They combines safety, quality and comfort, protecting against cuts, a common hazard in the meat and processing industries.
  • Detachable and interchangeable plastic straps with antimicrobial agent
  • Damaged straps can be changed over quickly and easily
  • Firm fastening for a secure fit every time; there is no side slit which may allow a knife to enter at wrist level
  • Fitted with an identification tag which can be personalised by the wearer
  • Each glove is
More Product Information