Disposal bin, SHARPSGUARD®, yellow
Supplier: DANIELS HEALTHCARE LIMITED
DD471YL DD473YL DD475YL DD472YL DD477YL DD442NRYL
129-1039EA 537 EUR
129-1039 129-1041 129-1043 129-1040 129-1063 DANIDD442NRYL
Disposal bin, SHARPSGUARD®, yellow
Bins Sharps Containers
SHARPSGUARD® yellow is a similar range of containers to SHARPSGUARD® orange, with the added advantage that they can also be used to dispose of sharps contaminated with medicinal products and their residues other than cytotoxic and/or cytostatic medicines. To ensure the containers are safe and simple to use the opaque doors clearly show when the door is in the temporary or fully closed position. The SHARPSGUARD® yellow range is designed to be incinerated or disposed of by other authorised process.
  • PP
The SHARPSGUARD® yellow theatre bin (129-1047) is designed to be used upright toMore Product Information