Sharps Containers, SHARPSGUARD® eco yellow
Supplier: DANIELS HEALTHCARE LIMITED
DD442NRYLR DD442YLR DD472YLR DD474YLR DD475YLR DD476YLR DD477YLR DD477WYLR DD509YLR DD440YLR DD439YLR DD471YLR
DANIDD442NRYLREA 159 EUR
DANIDD442NRYLR DANIDD442YLR DANIDD472YLR DANIDD474YLR DANIDD475YLR DANIDD476YLR DANIDD477YLR DANIDD477WYLR DANIDD509YLR DANIDD440YLR DANIDD439YLR DANIDD471YLR
Sharps Containers, SHARPSGUARD® eco yellow
Bins Sharps Containers
The SHARPSGUARD® eco yellow range is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic or cytostatic.
  • Sustainable alternative made from at least 40% recycled plastic
  • Small models can be easily fitted into a bag or glove compartment
  • Models eco yellow 7 and above are supplied with a robust pail handle for easy transportation
These containers are for disposal by incineration or other authorised process and designed for the consignment of waste categorised by the following EWC codes: 18 01 01, 18 01 03*, 18 01 09More Product Information