MF-Millipore™ Membrane Filters
Supplier: Merck
AAWP02500 AAWP03700 AAWP04700 GSWP02500 GSWP04700 HATF04700 HATF08250 HAWP03700 HAWP04700 PHWP04700 RAWG02500 RAWP01300 SMWP01300 SMWP04700 SSWP14250 VCWP04700 RAWP04700 AABP04700 AABG01300 AABG02500 AABG03700 AABG04700 AABP02500 AAWG01300 AAWG02500 AAWG0250C AAWG03700 AAWG04700 AAWP01300 AAWP037P0 AAWP037PM AAWP0470M AAWP09000 AAWP14250 DAWP01300 DAWP02500 DAWP09025 DAWP14250 GSTF01300 GSTF02500 GSTF09000 GSTF14250 GSWP03700 GSWP09000 HABG01300 HABG02500 HABG04700 HABP02500 HABP04700 HATF01300 HATF02500 HATF09025 HATF10050 HATF14250 HAWG01300 HAWG02500 HAWG03700 HAWG04700 HAWG05000 HAWP02400 HAWP0470M PHWP02500 PHWP09025 PHWP14250 HAWP09000 RATF14250 RAWG0250C RAWG04700 RAWP03700 SSWP01300 VSWP14250 VSWP04700 SCWP01300 SCWP02500 SCWP09025 SCWP14250 SMWP0190R SMWP02500 SMWP03700 SMWP09025 SMWP14250 SSWP02500 SSWP09025 VCWP01300 VCWP02500 VCWP09025 VCWP14250 VMWP01300 VMWP02500 VMWP04700 VMWP09025 VSWP09025 SSWP04700 SCWP04700 GSWP01300 VSWP02500 HAWP14250 GSTF04700 GSWP14250 RAWP02500 DAWP04700 RAWP14250 HAWP01300 VSWP01300 HAWP05000 HATF08550 HAWP02500 RAWP05000 RAWP09025 SCWP0190R HAEP04700 HAWG04705 AABPC5000 HAWP04700F1
AAWP02500EA 154 EUR
AAWP02500 AAWP03700 AAWP04700 GSWP02500 GSWP04700 HATF04700 HATF08250 HAWP03700 HAWP04700 PHWP04700 RAWG02500 RAWP01300 SMWP01300 SMWP04700 SSWP14250 VCWP04700 RAWP04700 AABP04700 AABG01300 AABG02500 AABG03700 AABG04700 AABP02500 AAWG01300 AAWG02500 AAWG0250C AAWG03700 AAWG04700 AAWP01300 AAWP037P0 AAWP037PM AAWP0470M AAWP09000 AAWP14250 DAWP01300 DAWP02500 DAWP09025 DAWP14250 GSTF01300 GSTF02500 GSTF09000 GSTF14250 GSWP03700 GSWP09000 HABG01300 HABG02500 HABG04700 HABP02500 HABP04700 HATF01300 HATF02500 HATF09025 MILFHATF10050 HATF14250 HAWG01300 HAWG02500 HAWG03700 HAWG04700 MILFHAWG05000 HAWP02400 HAWP0470M PHWP02500 PHWP09025 PHWP14250 HAWP09000 RATF14250 RAWG0250C RAWG04700 RAWP03700 SSWP01300 VSWP14250 VSWP04700 SCWP01300 SCWP02500 SCWP09025 SCWP14250 SMWP0190R SMWP02500 SMWP03700 SMWP09025 SMWP14250 SSWP02500 SSWP09025 VCWP01300 VCWP02500 VCWP09025 VCWP14250 VMWP01300 VMWP02500 VMWP04700 VMWP09025 VSWP09025 SSWP04700 SCWP04700 GSWP01300 VSWP02500 HAWP14250 GSTF04700 GSWP14250 RAWP02500 DAWP04700 RAWP14250 HAWP01300 VSWP01300 HAWP05000 HATF08550 HAWP02500 RAWP05000 RAWP09025 SCWP0190R HAEP04700 MILFHAWG04705 MILFAABPC5000 MILFHAWP04700F1
MF-Millipore™ Membrane Filters
Filters Membrane Filters
MF-Millipore™ filters without Triton® surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Biologically inert mixtures of cellulose acetate and cellulose nitrate.