Screw caps for tubes
Supplier: DELTALAB
617100 617200 617300 617000
DELT617100EA 319 EUR
DELT617100 DELT617200 DELT617300 DELT617000
Screw caps for tubes
Closures Test/Sample Tube Closures
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
  • Both models of caps are fully autoclavable and are pharmaceutical quality
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.