Tween® 20 (Polysorbate) 10% aqueous solution, Proteomics grade

Supplier: G Biosciences

Synonyms: Polyoxyethylene sorbitan monolaurate

DG011 DG012
DG011EA 327 EUR
DG011 DG012
Tween® 20 (Polysorbate) 10% aqueous solution, Proteomics grade
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR