ASB-14 ≥99%

Supplier: G Biosciences

Synonyms: Amidosulphobetaine-14

DG060 DG061
DG060EA 289 EUR
DG060 DG061
ASB-14 ≥99%
ASB-14

Zwitterionic detergent, Aminosulfobetaine with C14 alkyl tail is useful for solubilizing proteins for 2D analysis. Optimal solubility achieved in urea-thiourea mixtures and not in urea alone.


Reported to show better protein solubilization properties than CHAPS. ASB-14 has been shown to solubilise membrane proteins previously undetected.

Formula: C₂₂H₄₆N₂O₄S
CAS Number: 216667-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR