Oxivir Excel® CE Cleaner and Disinfectant
Supplier: Diversey
100941438
DIVY100941438EA 0 EUR
DIVY100941438
Oxivir Excel® CE Cleaner and Disinfectant
Cleaning Agents Disinfectants
Highly concentrated one step cleaner disinfectant with broad spectrum efficacy and short contact times. Based on Diversey Care patented Accelerated Hydrogen Peroxide formulation Oxivir Excel can be used on non invasive medical devices.
  • Full virucidal claim in just 30 seconds
  • Fast kill claims for a broad spectrum of viruses, bacteria and yeasts
  • Effective Cleaner
  • Fragrance free
  • No rinsing required 
  • Safe for the environment, the users and the patients, since the active ingredient breaks down into water and oxygen
  • Does not contain chlorine or alcohol