Disinfectants, ClearKlens Tego® 2000 VT25
Supplier: Diversey
7509461 100885537
DIVY7509461EA 449 EUR
DIVY7509461 141-1405
Disinfectants, ClearKlens Tego® 2000 VT25
Cleaning Agents Disinfectants
TEGO 2000 is a versatile and effective amphoteric-based disinfectant suitable for open plant cleaning applications throughout the food, beverage and dairy industries.
  • Ready to use, 0,2 micron filtered, sterile filled and double-bagged
  • Validated HPLC and titration test methods available
  • Compatible with ClearKlens Cleansinald and ClearKlens Bi-Spore as a rotational process
  • Highly effective terminal disinfectant for food industry use
  • Effective in soft or hard water
Tego 2000 should be applied after surfaces have been thoroughly cleaned and rinsed. It canMore Product Information