Supplier: DELTA T
A090007
DLTTA090007EA 111 EUR
DLTTA090007
Cooling Packs
37 C Accus maintain a temperature between 36 to 38 °C.
  • High Efficiency
  • Multi-use and disposable solution
  • Secured and safe
  • Control system
  • High availability
  • GDP, FDA, DQ, IQ, PQ, OQ compliance
Closure: aluminum composite foil and protective cap made of plastic
Color: PE natural
Fluid: red